Anti-CD34 Mouse Monoclonal Antibody [clone: SPM123]
Catalog # ANTIA250678-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD34
- Antigen symbol:CD34
- Clonality:Monoclonal
- Clone:SPM123
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Cynomolgus monkey,Rhesus monkey,Human
- Form:Liquid
- Gene ID:UniprotID# P28906
- Antigen synonyms:CD34 antigen|Cluster designation 34|Hematopoietic progenitor cell antigen CD34|CD34_HUMAN|Mucosialin|CD34 molecule|OTTHUMP00000034734|HPCA1|OTTHUMP00000034733
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:90 - 110 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Detergent solubilised vesicular suspension prepared from human term placenta
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [SPM123] antibody to CD34 for Flow Cytometry, IF and IHC-P with samples derived from Human, Cynomolgus Monkey and Rhesus Monkey.
- Validated applications: Flow cytometry, IF, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: SPM123
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Cynomolgus Monkey;Rhesus Monkey