Anti-PDLIM1 Mouse Monoclonal Antibody [clone: CPTC-PDLIM1-1]
ANTIA250480-100
New Product
- Antibody type:Primary
- Antigen name:C terminal LIM domain protein 1
- Antigen symbol:PDLIM1
- Clonality:Monoclonal
- Clone:CPTC-PDLIM1-1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O00151
- Antigen synonyms:Enigma homolog|Elfin|Enigma|CLIM 1|C-terminal LIM domain protein 1|CLP 36|Carboxy terminal LIM domain protein 1|CLIM1|CLP36
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:36 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human PDLIM1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [CPTC-PDLIM1-1] antibody to PDLIM1 for WB and IHC-P with samples derived from Human.
- Validated applications: WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: PDLIM1
Clonality: Monoclonal
Clone: CPTC-PDLIM1-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human