Anti-beta III Spectrin Mouse Monoclonal Antibody [clone: RG/26]
ANTIA253222-100
New Product
- Antibody type:Primary
- Antigen name:Beta III spectrin
- Antigen symbol:beta III Spectrin
- Clonality:Monoclonal
- Clone:RG/26
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O15020
- Antigen synonyms:Spectrin non erythroid beta chain 2|SCAR14|GTRAP41|Spectrin beta non erythrocytic 2|SCA5|Spectrin beta III sigma 2|SCA 5|Glutamate transporter EAAT4 associated protein 41|Spectrin beta chain brain 2
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:246 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Red cell membranes
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [RG/26] antibody to beta III Spectrin for ELISA, WB, IF and IHC-P with samples derived from Human, Mouse and Rat.
- Validated applications: ELISA, WB, IF, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: beta III Spectrin
Clonality: Monoclonal
Clone: RG/26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat