Anti-beta 2 Microglobulin Mouse Monoclonal Antibody [clone: B2M/961]
ANTIA252932-100
New Product
- Antibody type:Primary
- Antigen name:B2M
- Antigen symbol:beta 2 Microglobulin
- Clonality:Monoclonal
- Clone:B2M/961
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2b kappa
- Reactivity:Non-human primates,Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P61769
- Antigen synonyms:CDABP0092|Beta 2 microglobulin precursor |Hdcma22p|Beta chain of mhc class 1 proteins |Beta chain of MHC class I molecules|beta 2 Microglobulin|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin|B2MG_HUMAN
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:12 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human B2M protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [B2M/961] antibody to beta 2 Microglobulin for Flow Cytometry, IF, WB and IHC-P with samples derived from Human and Non-Human Primates.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 4 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: beta 2 Microglobulin
Clonality: Monoclonal
Clone: B2M/961
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Non-Human Primates