Order Entry
Export
ContactUsLinkComponent
Anti-beta 2 Microglobulin Mouse Monoclonal Antibody [clone: SPM617]
Anti-beta 2 Microglobulin Mouse Monoclonal Antibody [clone: SPM617]
Catalog # ANTIA252929-100
Supplier:  Antibodies.com
New Product
undefined
Anti-beta 2 Microglobulin Mouse Monoclonal Antibody [clone: SPM617]
Catalog # ANTIA252929-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    B2M
  • Antigen symbol:
    beta 2 Microglobulin
  • Clonality:
    Monoclonal
  • Clone:
    SPM617
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Non-human primates,
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P61769
  • Antigen synonyms:
    CDABP0092|Beta 2 microglobulin precursor |Hdcma22p|Beta chain of mhc class 1 proteins |Beta chain of MHC class I molecules|beta 2 Microglobulin|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin|B2MG_HUMAN
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    12 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human B2M protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [SPM617] antibody to beta 2 Microglobulin for WB, Flow Cytometry, IF and IHC-P with samples derived from Human and Non-Human Primates.

  • Validated applications: WB, Flow cytometry, IF, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: beta 2 Microglobulin
Clonality: Monoclonal
Clone: SPM617
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Non-Human Primates