Order Entry
Export
ContactUsLinkComponent
Anti-Cytokeratin 18 Mouse Monoclonal Antibody [clone: C-04]
Anti-Cytokeratin 18 Mouse Monoclonal Antibody [clone: C-04]
Catalog # ANTIA252382-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Cytokeratin 18 Mouse Monoclonal Antibody [clone: C-04]
Catalog # ANTIA252382-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cell proliferation inducing gene 46 protein
  • Antigen symbol:
    Cytokeratin 18
  • Clonality:
    Monoclonal
  • Clone:
    C-04
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Mouse,
    Cat,
    Marmoset,
    Hamster,
    Pig,
    Horse,
    Sheep,
    Goat,
    Ferret,
    Human,
    Bovine,
    Dog,
    Rat
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P05783
  • Antigen synonyms:
    CK18|Cell proliferation inducing protein 46|Cytokeratin 18|CYK18|Cytokeratin endo B|Cell proliferation-inducing gene 46 protein|CK 18|CK-18|CYK 18
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    45 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Cytoskeletal preparation extracted from the human vulvar squamous carcinoma cell line A431
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [C-04] antibody to Cytokeratin 18 for Flow Cytometry, IF, WB and IHC-P with samples derived from Human, Mouse, Rat, Hamster, Ferret, Bovine, Porcine, Feline, Canine, Sheep, Goat, Horse and Marmoset Monkey.

  • Validated applications: Flow cytometry, IF, WB, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cytokeratin 18
Clonality: Monoclonal
Clone: C-04
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Hamster, Ferret, Bovine, Porcine, Cat, Dog, Sheep, Goat, Horse, Marmoset Monkey