Anti-Integrin beta 3 Mouse Monoclonal Antibody [clone: Y2/51]
Catalog # ANTIA252259-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:BDPLT16
- Antigen symbol:Integrin beta 3
- Clonality:Monoclonal
- Clone:Y2/51
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P05106
- Antigen synonyms:HPA 1|CD61 antigen|GP3A|BDPLT2|CD 61|HPA 4|GPIIIa|GT|CD61
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:105/90 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Phytohemagglutinin (PHA)-stimulated peripheral blood cells
- Tested applications:IHC
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [Y2/51] antibody to Integrin beta 3 for Flow Cytometry, IF and IHC with samples derived from Human.
- Validated applications: Flow cytometry, IF, IHC
- Recommended dilutions: Flow cytometry: 0,5 to 1 µg/million cells, IF: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: Integrin beta 3
Clonality: Monoclonal
Clone: Y2/51
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human