Anti-ICAM3 Mouse Monoclonal Antibody [clone: ICO-60]
Catalog # ANTIA252088-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD50
- Antigen symbol:ICAM3
- Clonality:Monoclonal
- Clone:ICO-60
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P32942
- Antigen synonyms:ICAM 3|ICAM-3|CDW 50|ICAM3|Intercellular adhesion molecule 3|CDw50|ICAM3_HUMAN|ICAM-R|ICAMR
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:110 - 160 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human ICAM3 protein
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [ICO-60] antibody to ICAM3 for Flow Cytometry and IF with samples derived from Human.
- Validated applications: Flow cytometry, IF
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: ICO-60
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human