Order Entry
Export
ContactUsLinkComponent
Anti-HSP27 Mouse Monoclonal Antibody [clone: G3.1]
Anti-HSP27 Mouse Monoclonal Antibody [clone: G3.1]
Catalog # ANTIA252052-100
Supplier:  Antibodies.com
New Product
undefined
Anti-HSP27 Mouse Monoclonal Antibody [clone: G3.1]
Catalog # ANTIA252052-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    28 kDa heat shock protein
  • Antigen symbol:
    HSP27
  • Clonality:
    Monoclonal
  • Clone:
    G3.1
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Monkey,
    Sheep,
    Chicken,
    Chimpanzee,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P04792
  • Antigen synonyms:
    epididymis secretory protein Li 102|Heat shock 27 kDa protein|Estrogen regulated 24 kDa protein|Heat shock 27kD protein 1|Heat shock 27kDa protein|Estrogen-regulated 24 kDa protein|DKFZp586P1322|CMT2F|Heat shock 25kDa protein 1
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    27 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Partially purified HSP27 (originally called 24K) protein from breast cancer MCF-7 cells
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [G3.1] antibody to HSP27 for Flow Cytometry, IF, WB and IHC-P with samples derived from Human, Chimpanzee, Monkey, Sheep, Rat, Mouse and Chicken.

  • Validated applications: Flow cytometry, IF, WB, IHC-P
  • Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: HSP27
Clonality: Monoclonal
Clone: G3.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Chimpanzee, Monkey, Sheep, Rat, Mouse, Chicken