Anti-GDF8 Mouse Monoclonal Antibody [clone: GDF9/4261]
Catalog # ANTIA251884-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen symbol:GDF8
- Clonality:Monoclonal
- Clone:GDF9/4261
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O14793
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular weight:52 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:113 amino acid carboxy-terminal fragment of Myostatin protein
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [GDF9/4261] antibody to GDF8 for ELISA and WB with samples derived from Human.
- Validated applications: ELISA, WB
- Recommended dilutions: WB: 1 to 2 µg/ml
Type: Primary
Antigen: GDF8
Clonality: Monoclonal
Clone: GDF9/4261
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human