Order Entry
Export
ContactUsLinkComponent
Anti-Albumin Rabbit Recombinant Antibody [clone: ALB/6409R]
Anti-Albumin Rabbit Recombinant Antibody [clone: ALB/6409R]
Catalog # ANTIA251687-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Albumin Rabbit Recombinant Antibody [clone: ALB/6409R]
Catalog # ANTIA251687-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    alb
  • Antigen symbol:
    Albumin
  • Clonality:
    Recombinant
  • Clone:
    ALB/6409R
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P02768
  • Antigen synonyms:
    GIG20|DKFZp779N1935|ALBU_HUMAN|GIG42|Albumin (32 AA)|cell growth inhibiting protein 42|Albumin (AA 34)|Growth inhibiting protein 20|Albumin
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline; without Sodium Azide and carrier free
  • Molecular weight:
    66 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human ALB protein
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [ALB/6409R] antibody to Albumin for ELISA, WB and IHC-P with samples derived from Human.

  • Validated applications: ELISA, WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Albumin
Clonality: Recombinant
Clone: ALB/6409R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1 kappa
Reactivity: Human