Anti-CD47 Mouse Monoclonal Antibody [clone: CD47/3019] (Biotinylated)
Catalog # ANTIA251234-50
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Antigen identified by monoclonal
- Antigen symbol:CD47
- Clonality:Monoclonal
- Clone:CD47/3019
- Conjugation:Biotin
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q08722
- Antigen synonyms:IAP|CD47_HUMAN|CD47|CD 47|CD47 antigen|CD47 molecule|Antigenic surface determinant protein OA3|CD47 glycoprotein|CD47 antigen (Rh-related antigen, integrin-associated signal transducer)
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:~50 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:100 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, within amino acids 18 - 135, of human CD47 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:50 µG
Specifications
About this item
Mouse monoclonal [CD47/3019] antibody to CD47 (Biotin) for IF, Flow Cytometry and IHC-P with samples derived from Human.
- Validated applications: IF, Flow cytometry, IHC-P
- Recommended dilutions: IF: 2 to 4 µg/ml, Flow cytometry: 5 µl / million cells, IHC-P: 2 to 4 µg/ml
Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: CD47/3019
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human