Anti-CD34 Mouse Monoclonal Antibody [clone: ICO-115] (PE (Phycoerythrin))
Catalog # ANTIA251194-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD34
- Antigen symbol:CD34
- Clonality:Monoclonal
- Clone:ICO-115
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat
- Form:Liquid
- Gene ID:UniprotID# P28906
- Antigen synonyms:CD34 antigen|Cluster designation 34|Hematopoietic progenitor cell antigen CD34|CD34_HUMAN|Mucosialin|CD34 molecule|OTTHUMP00000034734|HPCA1|OTTHUMP00000034733
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:90 - 110 kDa
- Storage temperature:Store at 4...8 °C. Antibody is stable for 24 months.
- Concentration:100 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Blast cells of a chronic myeloid leukemia patient
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 T
Specifications
About this item
Mouse monoclonal [ICO-115] antibody to CD34 (PE) for Flow Cytometry and IF with samples derived from Human and Rat.
- Validated applications: Flow cytometry, IF
- Recommended dilutions: Flow cytometry: 5 to 10 µl / million cells, IF: 1:50 to 1:100
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: ICO-115
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat