Order Entry
Export
ContactUsLinkComponent
Anti-CD47 Mouse Monoclonal Antibody [clone: CD47/2937]
Anti-CD47 Mouse Monoclonal Antibody [clone: CD47/2937]
Catalog # ANTIA250731-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD47 Mouse Monoclonal Antibody [clone: CD47/2937]
Catalog # ANTIA250731-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Antigen identified by monoclonal
  • Antigen symbol:
    CD47
  • Clonality:
    Monoclonal
  • Clone:
    CD47/2937
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2c kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q08722
  • Antigen synonyms:
    IAP|CD47_HUMAN|CD47|CD 47|CD47 antigen|CD47 molecule|Antigenic surface determinant protein OA3|CD47 glycoprotein|CD47 antigen (Rh-related antigen, integrin-associated signal transducer)
  • Storage buffer:
    Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
  • Molecular weight:
    ~50 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, within amino acids 18 - 135, of human CD47 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [CD47/2937] antibody to CD47 for ELISA, WB and IHC-P with samples derived from Human.

  • Validated applications: ELISA, WB, IHC-P
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: CD47/2937
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c kappa
Reactivity: Human