Anti-CD44 Mouse Monoclonal Antibody [clone: DF1485]
Catalog # ANTIA250717-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:BA-1
- Antigen symbol:CD44
- Clonality:Monoclonal
- Clone:DF1485
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Mouse
- Form:Liquid
- Gene ID:UniprotID# P16070
- Antigen synonyms:CD44_HUMAN|CD44|CDw44|CD44 molecule (Indian blood group)|chondroitin sulfate proteoglycan 8|CD 44|Cell surface glycoprotein CD44|CD44 antigen|CD44 molecule
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:80 - 95 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Purified CD44 antigen (PGp-1) from lymphocyte membrane
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [DF1485] antibody to CD44 for Flow Cytometry, IF and IHC-P with samples derived from Human and Mouse.
- Validated applications: Flow cytometry, IF, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: DF1485
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse