Anti-CD44 Mouse Monoclonal Antibody [clone: SPM521]
Catalog # ANTIA250713-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:BA-1
- Antigen symbol:CD44
- Clonality:Monoclonal
- Clone:SPM521
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Baboon,Green monkey,Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P16070
- Antigen synonyms:CD44_HUMAN|CD44|CDw44|CD44 molecule (Indian blood group)|chondroitin sulfate proteoglycan 8|CD 44|Cell surface glycoprotein CD44|CD44 antigen|CD44 molecule
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:80 - 95 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Stimulated human leukocytes
- Tested applications:Functional studies, IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [SPM521] antibody to CD44 for IF, Functional Studies, WB and IHC-P with samples derived from Human, Baboon and Green Monkey.
- Validated applications: IF, Functional Studies, WB, IHC-P
- Recommended dilutions: IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 0,25 to 0,5 µg/ml
Type: Primary
Antigen: CD44
Clonality: Monoclonal
Clone: SPM521
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Baboon, Green Monkey