Anti-CD9 Mouse Monoclonal Antibody [clone: P1/33/2]
Catalog # ANTIA250571-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:5H9
- Antigen symbol:CD9
- Clonality:Monoclonal
- Clone:P1/33/2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P21926
- Antigen synonyms:Antigen defined by monoclonal|BTCC1|CD9 antigen p24|BA-2/p24 antigen|BTCC 1|BA2|CD9 antigen|5H9 antigen|CD9
- Storage buffer:Supplied in 10 mM Phosphate Buffered Saline with 0,05% BSA and 0,05% Sodium Azide
- Molecular weight:24 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human CD9 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [P1/33/2] antibody to CD9 for IHC-P with samples derived from Human.
- Validated applications: IHC-P
- Recommended dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: CD9
Clonality: Monoclonal
Clone: P1/33/2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human