Anti-Cytokeratin 20 Rabbit Polyclonal Antibody
Catalog # ANTIA12572-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD20
- Antigen symbol:Cytokeratin 20
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P35900
- Antigen synonyms:Cytokeratin-20|Keratin|CK-20|CK20|CK 20|Cytokeratin20|KA20|K1C20_HUMAN|Cytokeratin 20
- Storage buffer:Supplied in Phosphate buffered saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
- Molecular weight:43 kDa
- Sequence:KYEVMAQKNLQEAKEQFERQTAVLQQQVTVNTEELKGTEVQLTELRRTSQSLEIELQSHLSMKESLEHTLEETKARYSSQLANLQSLLSSLEAQLMQIRSNMERQNNEYHILLDIKTRLEQEIATYRRLLEGEDVKTTEYQLSTLEERDIKKTRKIKTVVQEVVDGKVVSSEVKEVEENI
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 245-424 of human Cytokeratin 20 (Cytokeratin 20 (KRT20)) (NP_061883.1)
- Tested applications:ICC/IF
- Purification:Affinity purification.
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to Cytokeratin 20 for WB and ICC/IF with samples derived from human and mouse.
- Validated applications: WB, ICC/IF
- Recommended dilutions: WB: 1:500 to 1:2000, ICC/IF: 1:50 to 1:20
Type: Primary
Antigen: Cytokeratin 20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse