Anti-Amyloid Precursor Protein Rabbit Polyclonal Antibody
Catalog # ANTIA12538-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:A4 amyloid protein
- Antigen symbol:Amyloid Precursor Protein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P05067
- Antigen synonyms:AICD-59|ABETA|AD1|A4_HUMAN|ABPP|AID(50)|AICD-50|AICD-57|AAA
- Storage buffer:Supplied in Phosphate buffered saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
- Molecular weight:110 kDa
- Sequence:AEESDNVDSADAEEDDSDVWWGGADTDYADGSEDKVVEVAEEEEVAEVEEEEADDDEDDEDGDEVEEEAEEPYEEATERTTSIATTTTTTTESVEEVVREV
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic peptide corresponding to a sequence within amino acids 190-290 of human APP (NP_000475.1)
- Tested applications:IHC
- Purification:Affinity purification.
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to Amyloid Precursor Protein for WB and IHC with samples derived from human.
- Validated applications: WB, IHC
- Recommended dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:20
Type: Primary
Antigen: Amyloid Precursor Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human