Order Entry
Export
ContactUsLinkComponent
Anti-SLC34A2 Rabbit Polyclonal Antibody
Anti-SLC34A2 Rabbit Polyclonal Antibody
Catalog # ANTIA12364-100
Supplier:  Antibodies.com
New Product
undefined
Anti-SLC34A2 Rabbit Polyclonal Antibody
Catalog # ANTIA12364-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Na(+)-dependent phosphate cotransporter 2B
  • Antigen symbol:
    SLC34A2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# O95436
  • Antigen synonyms:
    SLC34A2|NPT2B_HUMAN|NaPi-2b|Sodium-dependent phosphate transport protein 2B|Sodium-phosphate transport protein 2B|Solute carrier family 34 member 2|NaPi3b|Na(+)/Pi cotransporter 2B|Sodium/phosphate cotransporter 2B
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
  • Molecular weight:
    76 kDa
  • Sequence:
    EVATHYLEIITQLIVESFHFKNGEDAPDLLKVITKPFTKLIVQLDKKVISQIAMNDEKAKNKSLVKIWCKTFTNKTQINVTVPSTANCTSPSLCWTDGIQNWTMKNVTYKENIAKCQHIFVNFHLPDLA
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 234-362 of human SLC34A2 (NP_001171469.1)
  • Purification:
    Affinity purification.
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to SLC34A2 for WB with samples derived from human, mouse and rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:200

Type: Primary
Antigen: SLC34A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat