Order Entry
Export
ContactUsLinkComponent
Anti-CRY2 Rabbit Polyclonal Antibody
Anti-CRY2 Rabbit Polyclonal Antibody
Catalog # ANTIA12034-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CRY2 Rabbit Polyclonal Antibody
Catalog # ANTIA12034-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CRY2
  • Antigen symbol:
    CRY2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q49AN0
  • Antigen synonyms:
    FLJ10332|_HUMAN|KIAA0658|Cryptochrome 2|H|Cryptochrome-2|growth inhibiting protein 37|PHLL2|cryptochrome 2 (photolyase like)
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
  • Molecular weight:
    67 kDa
  • Sequence:
    MAATVATAAAVAPAPAPGTDSASSVHWFRKGLRLHDNPALLAAVRGARCVRCVYILDPWFAASSSVGINRWRFLLQSLEDLDTSLRKLNSRLFVVRGQPADVFPRLFKEWGVTRLTFEYDSEPFGKERDAAIMKMAKEAGVEVVTENSHTLYDLDRIIELNGQKPPLTYKRFQAIISRMELPKKPVGLVTSQQMESCRAEIQENHDETYGVPSLEELGFPTEGLGPAVWQ
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1 - 230 of human CRY2 (NP_066940.3)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to CRY2 for WB with samples derived from human and rat.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:1000

Type: Primary
Antigen: CRY2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat