Anti-BPI Rabbit Polyclonal Antibody
Catalog # ANTIA11825-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Bactericidal permeability-increasing protein
- Antigen symbol:BPI
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P17213
- Antigen synonyms:recombinant BPI holoprotein, rBPI|CAP 57|bactericidal/permeability-increasing protein|BPI|BPIFD1|rBPI|BPI fold containing family D, member 1
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
- Molecular weight:55 kDa
- Sequence:AETLDVQMKGEFYSENHHNPPPFAPPVMEFPAAHDRMVYLGLSDYFFNTAGLVYQEAGVLKMTLRDDMIPKESKFRLTTKFFGTFLPEVAKKFPNMKIQIHVSASTPPHLSVQPTGLTFYPAVDVQAFAVLPNSSLASLFLIGMHTTGSMEVSAESNRLVGELKLDRLLLELKHSNIGPFPVELLQDIMNYIVPILVLPRVNEKLQKGFPLPTPARVQLYNVVLQPHQNFLLFGADVVYK
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 248 - 487 of human BPI (NP_001716.2)
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody to BPI for WB with samples derived from human and mouse.
- Validated applications: WB
- Recommended dilutions: WB: 1:500 to 1:2000
Type: Primary
Antigen: BPI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse