Order Entry
Export
ContactUsLinkComponent
Anti-CD116 Rabbit Polyclonal Antibody
Anti-CD116 Rabbit Polyclonal Antibody
Catalog # ANTIA11504-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD116 Rabbit Polyclonal Antibody
Catalog # ANTIA11504-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD116
  • Antigen symbol:
    CD116
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P15509
  • Antigen synonyms:
    Colony stimulating factor 2 receptor alpha low affinity|CSF2RA|Colony stimulating factor 2 receptor alpha subunit|CD_antigen=|CDw116|antigen|CSF 2R|CSF2R|Colony stimulating factor 2 receptor alpha chain
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
  • Molecular weight:
    80 kDa
  • Sequence:
    EKSDLRTVAPASSLNVRFDSRTMNLSWDCQENTTFSKCFLTDKKNRVVEPRLSNNECSCTFREICLHEGVTFEVHVNTSQRGFQQKLLYPNSGREGTAAQNFSCFIYNADLMNCTWARGPTAPRDVQYFLYIRNSKRRREIRCPYYIQDSGTHVGCHLDNLSGLTSRNYFLVNGTSREIGIQFFDSLLDTKKIERFNPPSNVTVRCNTTHCLVRWKQPRTYQKLSYLDFQYQLDVHRKNTQPGTENLLINVSGDLENRYNFPSSEPRAKHSVKIRAADVRILNWSSWSEAIEFGSDDG
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 23 - 320 of human CSF2RA (NP_758448.1)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to CD116 for WB with samples derived from human and mouse.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: CD116
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse