Anti-beta 2 Microglobulin Rabbit Polyclonal Antibody
ANTIA11459-100
New Product
- Antibody type:Primary
- Antigen name:B2M
- Antigen symbol:beta 2 Microglobulin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P61769
- Antigen synonyms:CDABP0092|Hdcma22p|G_HUMAN|Beta chain of mhc class 1 proteins|Beta chain of MHC class I molecules|Beta 2 microglobulin precursor|beta 2 Microglobulin|Beta-2-microglobulin form pI 5.3|Beta 2 microglobin
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
- Molecular weight:12 kDa
- Sequence:IQRTPKIQVYSRHPAENGKSNFLNCYVSGFHPSDIEVDLLKNGERIEKVEHSDLSFSKDWSFYLLYYTEFTPTEKDEYACRVNHVTLSQPKIVKWDRDM
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 21 - 119 of human beta 2 Microglobulin (NP_004039.1)
- Tested applications:IHC
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Rabbit polyclonal antibody to beta 2 Microglobulin for WB and IHC with samples derived from human, mouse and rat.
- Validated applications: WB, IHC
- Recommended dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type: Primary
Antigen: beta 2 Microglobulin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat