Order Entry
Export
ContactUsLinkComponent
Anti-Cytokeratin 18 Rabbit Polyclonal Antibody
Anti-Cytokeratin 18 Rabbit Polyclonal Antibody
Catalog # ANTIA10906-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-Cytokeratin 18 Rabbit Polyclonal Antibody
Catalog # ANTIA10906-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cell proliferation inducing gene 46 protein
  • Antigen symbol:
    Cytokeratin 18
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P05783
  • Antigen synonyms:
    CK18|Cell proliferation inducing protein 46|Cytokeratin 18|CYK18|Cytokeratin endo B|Cell proliferation-inducing gene 46 protein|CK 18|CK-18|CYK 18
  • Modification:
    phospho Ser33
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
  • Molecular weight:
    48 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A phospho specific peptide corresponding to residues surrounding S33 of human KRT18
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to Cytokeratin 18 (phospho Ser33) for WB and IHC with samples derived from human.

  • Validated applications: WB, IHC
  • Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100

Type: Primary
Antigen: Cytokeratin 18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human