Order Entry
Export
ContactUsLinkComponent
Anti-FLVCR2 Rabbit Polyclonal Antibody
Anti-FLVCR2 Rabbit Polyclonal Antibody
Catalog # ANTIA10228-100
Supplier:  Antibodies.com
New Product
undefined
Anti-FLVCR2 Rabbit Polyclonal Antibody
Catalog # ANTIA10228-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Feline leukemia virus subgroup C cellular receptor family, member 2
  • Antigen symbol:
    FLVCR2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9UPI3
  • Antigen synonyms:
    FLVCR LIKE ON CHROMOSOME 14q|C14orf58|MFSD7C|FLVCR2|Calcium chelate transporter|CCT|FLVCRL14q|EPV|CHROMOSOME 14 OPEN READING FRAME 58
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
  • Molecular weight:
    57 kDa
  • Sequence:
    MVNEGPNQEESDDTPVPESALQADPSVSVHPSVSVHPSVSINPSVSVHPSSSAHPSALAQPSGLAHPSSSGPEDLSVIKVSRRRWAVVLVFSCYSMCNSF
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1 - 100 of human FLVCR2 (NP_060261.2)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to FLVCR2 for WB with samples derived from human and mouse.

  • Validated applications: WB
  • Recommended dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: FLVCR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse