Order Entry
Export
ContactUsLinkComponent
 
Anti-CD34 Mouse Monoclonal Antibody [clone: 581] (PE-Cyanine 5)
  ANTIA122058-100
New Product
Anti-CD34 Mouse Monoclonal Antibody [clone: 581] (PE-Cyanine 5)
  ANTIA122058-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    CD34
  • Antigen symbol:
    CD34
  • Clonality:
    Monoclonal
  • Clone:
    581
  • Conjugation:
    PE (Phycoerythrin)-Cy5®
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Non-human primates,
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P28906
  • Antigen synonyms:
    CD34 antigen|Cluster designation 34|Hematopoietic progenitor cell antigen CD34|CD34_HUMAN|Mucosialin|CD34 molecule|OTTHUMP00000034734|HPCA1|OTTHUMP00000034733
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Unfortunately, this information is considered to be co mMercially sensitive
  • Purification:
    This antibody is conjugated with tandem dye PE-Cyanine 5 under optimum conditions The conjugate is purified by size-exclusion chromatography
  • Pack type:
    Glass bottle
  • Pk:
    100 T

 

 

Mouse monoclonal [581] antibody to CD34 (PE-Cyanine 5) for Flow Cytometry with samples derived from human and Non-Human Primates.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 581
Conjugation: PE-Cyanine 5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Non-Human Primates