Anti-EGFR Mouse Monoclonal Antibody [clone: EGFR1] (PE-Cyanine 7)
Catalog # ANTIA122032-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Epidermal growth factor receptor
- Antigen symbol:EGFR
- Clonality:Monoclonal
- Clone:EGFR1
- Conjugation:PE (Phycoerythrin)-Cy7®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Horse,Human
- Form:Liquid
- Gene ID:UniprotID# P00533
- Antigen synonyms:EGFR_HUMAN|EGF R|EGFR|9030024J15Rik|Avian erythroblastic leukemia viral (verbb) oncogene homolog|Cell growth inhibiting protein 40|AI552599|Avian erythroblastic leukemia viral (v erb b) oncogene homolog|Cell proliferation inducing protein 61
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human epidermoid carcinoma line A431
- Purification:This antibody is conjugated with tandem dye PE-Cyanine 7 under optimum conditions The conjugate is purified by size-exclusion chromatography
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [EGFR1] antibody to EGFR (PE-Cyanine 7) for Flow Cytometry with samples derived from human and Horse.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: EGFR1
Conjugation: PE-Cyanine 7
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human;Horse