Anti-CD34 Mouse Monoclonal Antibody [clone: 581] (PerCP-Cyanine 5.5)
ANTIA121949-100
New Product
- Antibody type:Primary
- Antigen name:CD34
- Antigen symbol:CD34
- Clonality:Monoclonal
- Clone:581
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5,5®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Non-human primates,Human
- Form:Liquid
- Gene ID:UniprotID# P28906
- Antigen synonyms:CD34 antigen|Cluster designation 34|Hematopoietic progenitor cell antigen CD34|CD34_HUMAN|Mucosialin|CD34 molecule|OTTHUMP00000034734|HPCA1|OTTHUMP00000034733
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Unfortunately, this information is considered to be co mMercially sensitive
- Purification:This antibody is conjugated with tandem dye PerCP-Cyanine 55 under optimum conditions The conjugate is purified by size-exclusion chromatography
- Pack type:Glass bottle
- Pk:100 T
Mouse monoclonal [581] antibody to CD34 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from human and Non-Human Primates.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: 581
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Non-Human Primates