Order Entry
Export
ContactUsLinkComponent
Anti-CD19 Mouse Monoclonal Antibody [clone: 4G7] (PerCP-Cyanine 5.5)
Anti-CD19 Mouse Monoclonal Antibody [clone: 4G7] (PerCP-Cyanine 5.5)
Catalog # ANTIA121932-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD19 Mouse Monoclonal Antibody [clone: 4G7] (PerCP-Cyanine 5.5)
Catalog # ANTIA121932-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Antibody deficiency due to defect in CD19, included
  • Antigen symbol:
    CD19
  • Clonality:
    Monoclonal
  • Clone:
    4G7
  • Conjugation:
    PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5,5®
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P15391
  • Antigen synonyms:
    CD19 antigen|Cd19 protein|AW495831|B lymphocyte surface antigen B4|B-lymphocyte antigen CD19|B lymphocyte antigen CD19|CD19 molecule|CD19_HUMAN|B-lymphocyte surface antigen B4
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Human chronic lymphocytic leukemia (CCL) cells
  • Purification:
    This antibody is conjugated with tandem dye PerCP-Cyanine 55 under optimum conditions The conjugate is purified by size-exclusion chromatography
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [4G7] antibody to CD19 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: 4G7
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human