Order Entry
Export
ContactUsLinkComponent
Anti-CD151 Mouse Monoclonal Antibody [clone: 50-6] (PE (Phycoerythrin))
Anti-CD151 Mouse Monoclonal Antibody [clone: 50-6] (PE (Phycoerythrin))
Catalog # ANTIA121860-100
Supplier:  Antibodies.com
New Product
Anti-CD151 Mouse Monoclonal Antibody [clone: 50-6] (PE (Phycoerythrin))
Catalog # ANTIA121860-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD151
  • Antigen symbol:
    CD151
  • Clonality:
    Monoclonal
  • Clone:
    50-6
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P48509
  • Antigen synonyms:
    hemidesmosomal tetraspanin CD151|GP27|MER2|CD151 molecule|CD151_HUMAN|CD151 molecule (Raph blood group)|Membrane glycoprotein SFA-1|CD151 antigen|PETA-3
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Human epidermoid carcinoma cell line Hep-3
  • Purification:
    This antibody is conjugated with PE under optimum conditions The conjugate is purified by size-exclusion chromatography
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [50-6] antibody to CD151 (PE) for Flow Cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD151
Clonality: Monoclonal
Clone: 50-6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human