Order Entry
Export
ContactUsLinkComponent
Anti-MHC Class I Mouse Monoclonal Antibody [clone: IVA26]
Anti-MHC Class I Mouse Monoclonal Antibody [clone: IVA26]
Catalog # ANTIA121806-100
Supplier:  Antibodies.com
New Product
Anti-MHC Class I Mouse Monoclonal Antibody [clone: IVA26]
Catalog # ANTIA121806-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HLA class I histocompatibility antigen A 1 alpha chain
  • Antigen symbol:
    MHC Class I
  • Clonality:
    Monoclonal
  • Clone:
    IVA26
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Pig,
    Rabbit,
    Sheep,
    Coturnix japonica,
    Human,
    Bovine,
    Mouse
  • Form:
    Liquid
  • Antigen synonyms:
    H2 K1|Aw 68|Antigen presenting molecule|HLA A|H2K|CLASS I HISTOCOMPATIBILITY ANTIGEN|A 28|A 9|Aw 24
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Bovine lymphocytes
  • Tested applications:
    IHC-Fr
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [IVA26] antibody to MHC Class I for Flow Cytometry, IP and IHC-Fr with samples derived from mouse, Human, Porcine, Bovine, Rabbit, Sheep and Coturnix japonica.

  • Validated applications: Flow cytometry, IP, IHC to Fr
  • Recommended dilutions: Flow cytometry: 1 to 4 µg/ml

Type: Primary
Antigen: MHC Class I
Clonality: Monoclonal
Clone: IVA26
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Mouse;Human;Porcine;Bovine;Rabbit;Sheep;Coturnix japonica