Anti-KIR2DL4 Mouse Monoclonal Antibody [clone: mAb#33] (PE-Cyanine 5)
Catalog # ANTIA122047-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD158 antigen like family member D
- Antigen symbol:KIR2DL4
- Clonality:Monoclonal
- Clone:mAb#33
- Conjugation:PE (Phycoerythrin)/Cy5®
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:CD158d|KI2L4_HUMAN|CD158 antigen-like family member D|CD_antigen=CD158d|Killer cell inhibitory receptor 103AS|Killer cell immunoglobulin like receptor 2DL4|Killer cell immunoglobulin-like receptor 2DL4|Killer cell immunoglobulin like receptor, two domains, long cytoplasmic tail, 4|G9P
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:NK3.3 cells and KIR2DL4-Ig fusion protein
- Purification:This antibody is conjugated with tandem dye PE-Cyanine 5 under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [mAb#33] antibody to KIR2DL4 (PE-Cyanine 5) for Flow Cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: KIR2DL4
Clonality: Monoclonal
Clone: mAb#33
Conjugation: PE-Cyanine 5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human