Anti-CD45 Mouse Monoclonal Antibody [clone: 2D1] (PerCP-Cyanine 5.5)
Catalog # ANTIA121954-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:B220
- Antigen symbol:CD45
- Clonality:Monoclonal
- Clone:2D1
- Conjugation:PerCP-Cyanine 5.5
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P08575
- Antigen synonyms:CD45R|CD45|GP180|Leukocyte common antigen|L-CA|LCA|loc|CD45 antigen|CD 45
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human peripheral blood mononuclear cells
- Purification:This antibody is conjugated with tandem dye PerCP-Cyanine 5.5 under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [2D1] antibody to CD45 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD45
Clonality: Monoclonal
Clone: 2D1
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human