Anti-CD33 Mouse Monoclonal Antibody [clone: WM53] (PerCP-Cyanine 5.5)
Catalog # ANTIA121948-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 33
- Antigen symbol:CD33
- Clonality:Monoclonal
- Clone:WM53
- Conjugation:PerCP-Cyanine 5.5
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Non-human primates,Human
- Form:Liquid
- Gene ID:UniprotID# P20138
- Antigen synonyms:gp67|CD33|CD33 antigen|My9|CD33 antigen (gp67)|FLJ00391|CD33_HUMAN|CD33 molecule|Myeloid cell surface antigen CD33
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human AML cells
- Purification:This antibody is conjugated with tandem dye PerCP-Cyanine 5.5 under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [WM53] antibody to CD33 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from human and Non-Human Primates.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD33
Clonality: Monoclonal
Clone: WM53
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-Human Primates