Anti-CD3 Mouse Monoclonal Antibody [clone: OKT3] (PerCP-Cyanine 5.5)
Catalog # ANTIA121944-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:4930549J05Rik
- Antigen symbol:CD3
- Clonality:Monoclonal
- Clone:OKT3
- Conjugation:PerCP-Cyanine 5.5
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P07766
- Antigen synonyms:CD247 antigen|CD3|CD3 antigen, delta subunit|CD247 antigen, zeta subunit|CD247 molecule|Cd247|CD3 delta|A430104F18Rik|AW552088
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Unfortunately, this information is considered to be commercially sensitive
- Purification:This antibody is conjugated with tandem dye PerCP-Cyanine 5.5 under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [OKT3] antibody to CD3 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD3
Clonality: Monoclonal
Clone: OKT3
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human