Anti-CD27 Mouse Monoclonal Antibody [clone: LT27] (PerCP-Cyanine 5.5)
ANTIA121938-100
New Product
- Antibody type:Primary
- Antigen name:CD 27
- Antigen symbol:CD27
- Clonality:Monoclonal
- Clone:LT27
- Conjugation:PerCP-Cyanine 5.5
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P26842
- Antigen synonyms:CD27_HUMAN|S152|CD27 antigen|CD27 molecule|CD27L receptor|MGC20393|OTTHUMP00000238557|CD27|LPFS2
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human peripheral blood lymphocytes
- Purification:This antibody is conjugated with tandem dye PerCP-Cyanine 5.5 under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Mouse monoclonal [LT27] antibody to CD27 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD27
Clonality: Monoclonal
Clone: LT27
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human