Order Entry
Export
ContactUsLinkComponent
Anti-CD24 Mouse Monoclonal Antibody [clone: SN3] (PerCP-Cyanine 5.5)
Anti-CD24 Mouse Monoclonal Antibody [clone: SN3] (PerCP-Cyanine 5.5)
Catalog # ANTIA121936-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD24 Mouse Monoclonal Antibody [clone: SN3] (PerCP-Cyanine 5.5)
Catalog # ANTIA121936-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD 24
  • Antigen symbol:
    CD24
  • Clonality:
    Monoclonal
  • Clone:
    SN3
  • Conjugation:
    PerCP-Cyanine 5.5
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P25063
  • Antigen synonyms:
    CD24 antigen (small cell lung carcinoma cluster 4 antigen)|CD24A|FLJ43543|CD24|CD24 antigen|FLJ22950|CD24_HUMAN|CD24 molecule|GPI linked surface mucin
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Glycoproteins purified from human NALM-1 cell line
  • Purification:
    This antibody is conjugated with tandem dye PerCP-Cyanine 5.5 under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [SN3] antibody to CD24 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.

Type: Primary
Antigen: CD24
Clonality: Monoclonal
Clone: SN3
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human