Anti-CD20 Mouse Monoclonal Antibody [clone: 2H7] (PerCP-Cyanine 5.5)
Catalog # ANTIA121933-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:APY
- Antigen symbol:CD20
- Clonality:Monoclonal
- Clone:2H7
- Conjugation:PerCP-Cyanine 5.5
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Reactivity:Non-human primates,Human
- Form:Liquid
- Gene ID:UniprotID# P11836
- Antigen synonyms:B-lymphocyte surface antigen B1|B Lymphocyte Cell Surface Antigen B1|Bp35|ATOPY|CD 20|B-lymphocyte antigen CD20|B-lymphocyte cell-surface antigen B1|Bp 35|B lymphocyte antigen CD20
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Human tonsillar B cells
- Purification:This antibody is conjugated with tandem dye PerCP-Cyanine 5.5 under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [2H7] antibody to CD20 (PerCP-Cyanine 5.5) for Flow Cytometry with samples derived from human and Non-Human Primates.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 4 µl reagent / 100 µl of whole blood or 106 cells in a suspension.
Type: Primary
Antigen: CD20
Clonality: Monoclonal
Clone: 2H7
Conjugation: PerCP-Cyanine 5.5
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human, Non-Human Primates