Anti-MEK1 + MEK2 Rabbit Polyclonal Antibody
ANTIA121815-50
New Product
- Antibody type:Primary
- Antigen name:AA589381
- Antigen symbol:MEK1 + MEK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:MAP kinase kinase 2|Dual specificity mitogen-activated protein kinase kinase 1|CFC3|ERK activator kinase 2|ERK activator kinase 1|EC 2.7.12.2|Dual specificity mitogen-activated protein kinase kinase 2|FLJ26075|MAP kinase kinase 1
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide corresponding to amino acids 33-53 of human MAPK Kinase 1, coupled with KLH
- Purification:Affinity chromatography
- Pack type:Plastic vial
- Pk:50 µG
Rabbit polyclonal antibody to MEK1 + MEK2 for WB with samples derived from human.
- Validated applications: WB
- Recommended dilutions: WB: 0,5 µg/ml
Type: Primary
Antigen: MEK1 + MEK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human