Anti-IL-18 Mouse Monoclonal Antibody [clone: IL18/7781]
Catalog # ANTIA316534-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Iboctadekin
- Antigen symbol:IL-18
- Clonality:Monoclonal
- Clone:IL18/7781
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q14116
- Antigen synonyms:IFN gamma inducing factor|IL-1 gamma|IL 1g|IL 18|IFN-gamma-inducing factor|IGIF|IL1 gamma|IL-18|IL 1 gamma
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:18 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-193) of human IL-18 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [IL18/7781] antibody to IL-18 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: IL-18
Clonality: Monoclonal
Clone: IL18/7781
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2 kappa
Reactivity: Human