Anti-c-Myc Rabbit Recombinant Antibody [clone: MYC/7150R]
Catalog # ANTIA316185-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:AU016757
- Antigen symbol:c-Myc
- Clonality:Recombinant
- Clone:MYC/7150R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P01106
- Antigen synonyms:MRTL|Class E basic helix-loop-helix protein 39|bHLHe39|c Myc|Avian myelocytomatosis viral oncogene homolog|c-Myc|Myc Epitope Tag|Myc protein|Myc proto oncogene protein
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:62 - 64 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 1-200) of human MYC protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [MYC/7150R] antibody to c-Myc for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: c-Myc
Clonality: Recombinant
Clone: MYC/7150R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human