Anti-CD21 Rabbit Recombinant Antibody [clone: CR2/7185R]
Catalog # ANTIA316005-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:C3DR
- Antigen symbol:CD21
- Clonality:Recombinant
- Clone:CR2/7185R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P20023
- Antigen synonyms:Complement C3d receptor|CR|Complement component (3d/Epstein Barr virus) receptor 2|CD 21|Complement C3d receptor 2|Complement receptor type 2|CD21|Cr2|CR2_HUMAN
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:140 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment corresponding to C-terminal of human CD21. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [CR2/7185R] antibody to CD21 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: CD21
Clonality: Recombinant
Clone: CR2/7185R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human