Anti-CD20 Mouse Recombinant Antibody [clone: rMS4A1/8044]
ANTIA316000-100
New Product
- Antibody type:Primary
- Antigen name:APY
- Antigen symbol:CD20
- Clonality:Recombinant
- Clone:rMS4A1/8044
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P11836
- Antigen synonyms:B-lymphocyte surface antigen B1|B Lymphocyte Cell Surface Antigen B1|Bp35|ATOPY|CD 20|B-lymphocyte antigen CD20|B-lymphocyte cell-surface antigen B1|Bp 35|B lymphocyte antigen CD20
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:33 - 37 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at +4 °C
- Immunogen:Recombinant fragment (around Amino Acids 200-297) of human MS4A1 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant mouse monoclonal [rMS4A1/8044] antibody to CD20 for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: CD20
Clonality: Recombinant
Clone: rMS4A1/8044
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human