Anti-MZF1 Mouse Monoclonal Antibody [clone: PCRP-MZF1-1E8]
Catalog # ANTIA315358-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Myeloid zinc finger 1
- Antigen symbol:MZF1
- Clonality:Monoclonal
- Clone:PCRP-MZF1-1E8
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P28698
- Antigen synonyms:Zfp98|Zinc finger and SCAN domain containing protein 6|MZF1|MZF1_HUMAN|MZF1B|Zinc finger protein 42|MZF 1|Zinc finger and SCAN domain-containing protein 6|MZF-1
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
- Molecular weight:82,05 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment (around Amino Acids 37-128) of human MZF1 protein
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [PCRP-MZF1-1E8] antibody to MZF1 for flow cytometry, IF and IHC-P with samples derived from human.
- Validated applications: Flow cytometry, IF, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: MZF1
Clonality: Monoclonal
Clone: PCRP-MZF1-1E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human