Order Entry
Export
ContactUsLinkComponent
Anti-ICAM1 Rabbit Recombinant Antibody [clone: ICAM1/8677R]
Anti-ICAM1 Rabbit Recombinant Antibody [clone: ICAM1/8677R]
Catalog # ANTIA315124-100
Supplier:  Antibodies.com
New Product
Anti-ICAM1 Rabbit Recombinant Antibody [clone: ICAM1/8677R]
Catalog # ANTIA315124-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Antigen identified by monoclonal
  • Antigen symbol:
    ICAM1
  • Clonality:
    Recombinant
  • Clone:
    ICAM1/8677R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P05362
  • Antigen synonyms:
    Human rhinovirus receptor|CD54 antigen|Cell surface glycoprotein P3.58|BB2|CD54|CD_antigen=CD54|BB 2|CD 54|ICAM 1
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide.
  • Molecular weight:
    85 - 115 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment raised against intracellular epitope of human ICAM1. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [ICAM1/8677R] antibody to ICAM1 for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ml

Type: Primary
Antigen: ICAM1
Clonality: Recombinant
Clone: ICAM1/8677R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human