Order Entry
Export
ContactUsLinkComponent
Anti-Cytokeratin 8 + 18 Mouse Monoclonal Antibody [clone: 5D3]
Anti-Cytokeratin 8 + 18 Mouse Monoclonal Antibody [clone: 5D3]
Supplier:  ANTIBODIES.COM
New Product
Certificates
undefined
Anti-Cytokeratin 8 + 18 Mouse Monoclonal Antibody [clone: 5D3]
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Catalog No:
  • ANTIA254291-500
  • ANTIA254291-100
  • Antigen symbol:
  • Cytokeratin 8 + 18
  • Cytokeratin 8 + 18
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 5D3
  • 5D3
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • UniprotID# P05787, P05783
  • UniprotID# P05787, P05783
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Cytoskeleton preparation from human breast cancer MCF-7 cells
  • Cytoskeleton preparation from human breast cancer MCF-7 cells
  • Form:
  • Liquid
  • Liquid
  • Molecular weight:
  • 52,5 kDa (CK8)/45 kDa (CK18)
  • 52,5 kDa (CK8)/45 kDa (CK18)
  • Storage buffer:
  • Supplied as an aliquot of tissue culture supernatant with 0,05% sodium azide
  • Supplied as an aliquot of tissue culture supernatant with 0,05% sodium azide
  • Storage temperature:
  • Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C
  • Tested applications:
  • IHC-P
  • IHC-P

Specifications