Anti-beta III Spectrin Mouse Monoclonal Antibody [clone: SPTBN2/1584]
Catalog # ANTIA250046-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Beta III spectrin
- Clonality:Monoclonal
- Clone:SPTBN2/1584
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O15020
- Antigen synonyms:Spectrin non erythroid beta chain 2|SCAR14|GTRAP41|Spectrin beta non erythrocytic 2|SCA5|Spectrin beta III sigma 2|SCA 5|Glutamate transporter EAAT4 associated protein 41|Spectrin beta chain brain 2
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:246 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 356-475, of human SPTBN2 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [SPTBN2/1584] antibody to beta III Spectrin for flow cytometry, IF, WB and IHC-P with samples derived from human, rat and mouse.
- Validated applications: Flow cytometry, IF, WB, IHC-P
- Recommended dilutions: Flow cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: beta III Spectrin
Clonality: Monoclonal
Clone: SPTBN2/1584
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Rat, Mouse