Anti-Cytokeratin 20 Mouse Monoclonal Antibody [clone: SPM140]
ANTIA249671-100
New Product
- Antibody type:Primary
- Antigen name:CD20
- Antigen symbol:Cytokeratin 20
- Clonality:Monoclonal
- Clone:SPM140
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2b kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P35900
- Antigen synonyms:Cytokeratin-20|Keratin|CK-20|CK20|CK 20|Cytokeratin20|KA20|K1C20_HUMAN|Cytokeratin 20
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:46 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 196-323, of human KRT20 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [SPM140] antibody to Cytokeratin 20 for WB and IHC-P with samples derived from human.
- Validated applications: WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 20
Clonality: Monoclonal
Clone: SPM140
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human