Order Entry
Export
ContactUsLinkComponent
Anti-n-Myc Mouse Monoclonal Antibody [clone: NMYC-1]
Anti-n-Myc Mouse Monoclonal Antibody [clone: NMYC-1]
Catalog # ANTIA249458-100
Supplier:  Antibodies.com
New Product
undefined
Anti-n-Myc Mouse Monoclonal Antibody [clone: NMYC-1]
Catalog # ANTIA249458-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    bHLHe37
  • Antigen symbol:
    n-Myc
  • Clonality:
    Monoclonal
  • Clone:
    NMYC-1
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P04198
  • Antigen synonyms:
    N myc|mycn|n-Myc|Class E basic helix-loop-helix protein 37|N-myc proto-oncogene protein|Neuroblastoma derived v myc avian myelocytomatosis viral related oncogene|MODED|MYCN_HUMAN|N myc proto oncogene protein
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    67 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human n-Myc protein
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [NMYC-1] antibody to n-Myc for IF and WB with samples derived from human, mouse and rat.

  • Validated applications: IF, WB
  • Recommended dilutions: IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml

Type: Primary
Antigen: n-Myc
Clonality: Monoclonal
Clone: NMYC-1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat